Learn More
Adenovirus Type 5 Hexon Polyclonal Antibody, FITC, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA173053
Code : LD
Additional Details : Weight : 0.01000kg
Description
The PA1-73053 antibody reacts with Adenovirus types 2-6. This antibody does not cross-react with Para 1-3, Influenza A &B, RSV, or hMPV. This antibody is negative against uninfected Hep-2, LLCMK2, and Vero cells by direct IF. PA1-73053 has been successfully used in immunofluorescence applications. Direct immunofluorescence was performed on an acetone-fixed permissive tissue culture system The PA1-73053 antibody was raised against gradient purified, disrupted Adenovirus type 5 virions. Recommended short term storage of less than 6 months at 4oC under subdued light. For long term storage, aliquot and store at -20oC.
Adenovirus Type 5 Hexon Polyclonal antibody specifically detects Adenovirus Type 5 Hexon in Virus samples. It is validated for Immunocytochemistry.Specifications
Adenovirus Type 5 Hexon | |
Polyclonal | |
FITC | |
Adeno Virus; Adeno Virus 5; AdV; AdV-5; HADV; HADV-5 | |
Gradient purified, disrupted Adenovirus type 5 virions. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2 | |
Goat | |
purified | |
RUO | |
Virus | |
Antibody | |
IgG |