missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenovirus Type 2 Fiber Monoclonal Antibody (2A6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA134961
Code : LE
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Adenovirus infected cells and tissues..
Adenovirus Type 2 Fiber Monoclonal antibody specifically detects Adenovirus Type 2 Fiber in Virus samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Adenovirus Type 2 Fiber | |
Monoclonal | |
0.2 mg/mL | |
10mM PBS with 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry, Western Blot | |
2A6 | |
Unconjugated | |
Adeno Virus; Adeno Virus 2; AdV; AdV-2; HADV; HADV-2 | |
UV irradiated Ad2 virus. | |
500 μL | |
Primary | |
4° C, do not freeze | |
Liquid |