Learn More
Adenovirus Type 7 Monoclonal Antibody (1N14), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA17600
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-7600 detects Adenovirus from virus samples. MA1-7600 has been successfully used in ELISA, Western blot and Immunofluorescence procedures. The MA1-7600 immunogen is a purified Ad7 virus.
Adenovirus Type 7 Monoclonal antibody specifically detects Adenovirus Type 7 in Virus samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Adenovirus Type 7 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.01% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
ELISA, Immunocytochemistry, Western Blot | |
1N14 | |
Unconjugated | |
Adeno Virus; Adeno Virus 7; AdV; AdV-7; HADV; HADV-7 | |
Purified Ad7 virus. | |
100 μg | |
Primary | |
4°C short term, -80°C long term | |
Liquid |