Learn More
AAV4 Virus VP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14107
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-4107 detects adeno-associated 4 virus. PA1-4107 has been successfully used in Western blot procedures. The PA1-4107 immunogen is a synthetic peptide corresponding to residues N(456) F T K L R P T N F S N C(467) of the capsid VP3 protein of AAV 4.
AAV4 Virus VP3 Polyclonal antibody specifically detects AAV4 Virus VP3 in Virus samples. It is validated for Western Blot.Specifications
AAV4 Virus VP3 | |
Polyclonal | |
Unconjugated | |
AAV; AAV-4 VP3; AdV4; AV4; HADV-4; Lentivirus | |
Synthetic peptide corresponding to residues N(456) F T K L R P T N F S N C(467) of the capsid VP3 protein of AAV 4. | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Rabbit | |
Affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |