Learn More
ADAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531451
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG, MCF-7. Predicted reactivity: Mouse (85%), Rat (85%), Rhesus Monkey (97%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ADAT1 Polyclonal antibody specifically detects ADAT1 in Human samples. It is validated for Western Blot.Specifications
ADAT1 | |
Polyclonal | |
Unconjugated | |
ADAT1 | |
Adat1; Adenosine deaminase; adenosine deaminase acting on tRNA; adenosine deaminase tRNA specific 1; adenosine deaminase, tRNA specific 1; adenosine deaminase, tRNA-specific 1; HADAT1; mADAT1; MMADAT1; tRNA-specific 1; tRNA-specific adenosine deaminase 1; tRNA-specific adenosine-37 deaminase; zgc:162299 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23536 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9BUB4 | |
ADAT1 | |
Recombinant fragment corresponding to a region within amino acids 209 and 477 of Human ADAT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |