Learn More
ADAM32 Mouse anti-Human, Clone: 539220, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5380
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460926
  • £249.49 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
ADAM32 Monoclonal antibody MAB5380 Specifically detects ADAM32 in Human samples. It is validated for Immunocytochemistry
Specifications
Specifications
ADAM32
539220
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ADAM32
Chinese hamster ovary cell line CHO-derived recombinant human ADAM32 Ser17-Thr476 Accession # Q8TC27
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
203102
Reconstitute at 0.5 mg/mL in sterile PBS.
Immunocytochemistry
LYOPH
Immunocytochemistry 8-25 ug/mL
Q8TC27
a disintegrin and metalloprotease domain 32, a disintegrin and metalloproteinase domain 32, ADAM 32, ADAM metallopeptidase domain 32, disintegrin and metalloproteinase domain-containing protein 32, FLJ26299, FLJ29004, metalloproteinase 12-like protein
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human ADAM32 in direct ELISAs.