missing translation for 'onlineSavingsMsg'
Learn More

ADAM19 Ectodomain Rat anti-Mouse, Clone: 164646, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1229

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488465

  • £353.77 / 500µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

ADAM19 Ectodomain Monoclonal antibody MAB1229 Specifically detects ADAM19 Ectodomain in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

ADAM19
164646
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ADAM19
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse ADAM19 Ectodomain, Asp205-Gly705 (predicted), Accession # O35674
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
8728
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O35674
a disintegrin and metalloproteinase domain 19 (meltrin beta), ADAM 19, ADAM metallopeptidase domain 19, disintegrin and metalloproteinase domain-containing protein 19, EC 3.4.24, EC 3.4.24.-, MADDAM, Meltrin beta, Meltrin-beta, Metalloprotease and disintegrin dendritic antigen marker, MLTNBmeltrin beta
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse ADAM19 Ectodomain in direct ELISAs and Western blots. It also recognizes a C-terminal fragment (amino acids 588-705) of recombinant mouse (rm) ADAM19 in Western blots. In Western blots, no cross-reactivity with rmADAM10 or rmADAM15 is observed.
Mouse
Videos

For Research Use Only