missing translation for 'onlineSavingsMsg'
Learn More

Acinus Polyclonal Antibody, Invitrogen™

Acinus Antibody, PA1-12497, from Invitrogen™. Species Reactivity: Human; Applications: Western Blot.

Brand:  Invitrogen PA112497

UNSPSC : 12352203

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11525982

  • £292.00 / 100µg


Estimated Shipment
19-10-2017
Add to basket

Description

Description

Chromatin condensation and nuclear fragmentation (CCNF)is the hallmark of apoptosis. CCNF is triggered by the activation of members of caspase family, caspase activated DNase (CAD/DFF40), and several novel proteins including AIF and CIDE. A new inducer of chromatin condensation was recently identified and designated Acinus (for apoptotic chromatin condensation inducer in the nucleus). Acinus is cleaved by caspase-3 and an additional unknown protease generating a small active peptide p17, which causes chromatin condensation in vitro when it is added to purified nuclei. Acinus also induces apoptotic chromatin condensation in cells. Acinus is ubiquitously expressed.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic peptide (SRSTPVRDRGGRR) corresponding to aa 1329-1341 of human AcinusL, 602-614 of human AcinusS' or 571-583 of human AcinusS
Formulation: 100μg purified antibody (0.5mg/mL) in PBS and 0.02% sodium azide
Synonyms: Apoptotic chromatin condensation inducer in the nucleus
Uniprot Accession: Q9UKV3
Entrez GeneID: 22985
Gene Symbol: ACIN1
A suggested positive control for this product is K562 cell lysate.
Specifications

Specifications

Acinus
Unconjugated
Q9UKV3
Apoptotic chromatin condensation inducer in the nucleus
Rabbit
IgG
Avoid Freeze/Thaw Cycles
Polyclonal
Human
Western Blot
100μg purified antibody (0.5mg/mL) in PBS and 0.02% sodium azide
Purified
ACIN1
602-614 of human AcinusS' or 571-583 of human AcinusS
100μg
Primary
22985