missing translation for 'onlineSavingsMsg'
Learn More

Acinus, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA112496

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11515982

  • £260.94 / 100µg


Estimated Shipment
29-05-2018
Add to basket

Description

Description

A suggested positive control for this product is K562 cell lysate.
Specifications

Specifications

Acinus
Polyclonal
PBS with 0.02% sodium azide
Q9UKV3
Apoptotic chromatin condensation inducer in the nucleus
Rabbit
IgG
100μg
Antibody
Polyclonal
Human
Western Blot
Unconjugated
ACIN1
Purified
ACIN1
Synthetic peptide (TRTALHGVKWPQSNPK) corresponding to aa 1065-1080 of human AcinusL, 338-353 of human AcinusS' or 307-322 of human AcinusS. Sequence is located near the C-terminus of the cleaved active peptide p17.
Antigen affinity chromatography
Avoid Freeze/Thaw Cycles, store at -20°C
Primary
22985
Videos