Learn More
ACE/CD143 Mouse anti-Human, Clone: 171409, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB9291
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13442006
  • £337.93 / 500µg
Estimated Shipment
08-07-2019
Description
Description
ACE/CD143 Monoclonal antibody MAB9291 Specifically detects ACE/CD143 in Human samples. It is validated for Western Blotting
Specifications
Specifications
ACE/CD143
171409
Western Blot 1 ug/mL
P12821.1
ACE1angiotensin converting enzyme, somatic isoform, angiotensin I converting enzyme (peptidyl-dipeptidase A) 1, carboxycathepsin, CD143, CD143 antigen, DCP, DCP1, DCP1angiotensin-converting enzyme, dipeptidyl carboxypeptidase 1, Dipeptidyl carboxypeptidase I, EC 3.2.1.-, EC 3.4.15.1, Kininase II, MGC26566, MVCD3, peptidase P, testicular ECA
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects recombinant human (rh)ACE in direct ELISAs and Western blots. Does not cross-react with rhACE-2 or rhNeprilysin.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ACE
Mouse myeloma cell line NS0-derived recombinant human ACE Leu30-Leu1261 Accession # P12821.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1636
Reconstitute at 0.5 mg/mL in sterile PBS.