Learn More
4-1BB Ligand/TNFSF9 Rat anti-Mouse, Clone: 203942, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1246
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488485
  • £397.00 / 500µg
Estimated Shipment
28-06-2018
Add to basket
Description
Description
4-1BB Ligand/TNFSF9 Monoclonal antibody MAB1246 Specifically detects 4-1BB Ligand/TNFSF9 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
4-1BB Ligand/TNFSF9
203942
Purified
S. frugiperda insect ovarian cell linesf 21-derived recombinant mouse 4-1BB Ligand/TNFSF9. Arg104-Glu309
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
CyTOF
Unconjugated
Rat
IgG2b
500μg
Primary
Murine