Learn More
14-3-3 beta/epsilon/zeta Monoclonal Antibody (3C8), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA134561
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-34561 detects 14-3-3 beta + epsilon + zeta from human samples. MA1-34561 has been successfully used in Western blot applications. The MA1-34561 immunogen is: Human 14-3-3 beta fusion protein.
Specifications
14-3-3 beta/epsilon/zeta | |
Monoclonal | |
1 mg/mL | |
YWHAE | |
YWHAB, YWHAE, YWHAZ, 14-3-3 protein beta/alpha, KCIP-1, Protein kinase C inhibitor protein 1, Protein 1054, 14-3-3 protein epsilon, 14-3-3e, 14-3-3 protein zeta, 14-3-3 protein delta, 14-3-3z, 14-3-3d | |
Mouse | |
Protein A | |
RUO | |
22627, 22631, 25578, 29753, 54401, 56011, 7529, 7531, 7534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
3C8 | |
Unconjugated | |
P31946, P62258, P63104, P62259, P63101, Q9CQV8, P35213, P62260, P63102 | |
YWHAB, YWHAE, YWHAZ | |
Human 14-3-3 beta fusion protein. | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |