missing translation for 'onlineSavingsMsg'
Learn More

Anthrone, ACS reagent, Thermo Scientific Chemicals

Catalog Number p-3735533
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
90-44-8
C14H10O
194.23
MFCD00001187
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
7018
CHEBI:33835
10H-anthracen-9-one
O=C1C2=CC=CC=C2CC2=CC=CC=C12
90-44-8
C14H10O
194.23
MFCD00001187
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
anthrone, 9 10h-anthracenone, carbothrone, anthranone, 9-oxoanthracene, 9,10-dihydro-9-oxoanthracene, az-o, anthracen-9 10h-one, anthracene, 9,10-dihydro-9-oxo, unii-fp0fj7k744
7018
CHEBI:33835
10H-anthracen-9-one
O=C1C2=CC=CC=C2CC2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Absorbance 0.75 max. (425nm, 1cm cell) Passes Test, 0.045 max. (620nm, 1cm cell) Passes Test
Melting Point 152.0°C to 157.0°C
Color Yellow
Boiling Point 721.0°C
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 25 g
Beilstein 07,473
Sensitivity (to carbohydrates) Passes Test
Merck Index 15,682
Solubility Information (in ethyl acetate) Passes test
Formula Weight 194.23
Grade ACS Reagent
Physical Form Crystals
Chemical Name or Material Anthrone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'