missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anthranilamide, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 111.0°C to 114.0°C |
Boiling Point | 300.0°C |
Flash Point | 198°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (HPLC) |
Packaging | Plastic bottle |
Linear Formula | 2-(H2N)C6H4CONH2 |
Quantity | 500 g |
Beilstein | 14, 320 |
Solubility Information | Solubility in water: <5g/L water (20°C). Other solubilities: 140g/L ethanol (20°C), slightly soluble in ethylether, benzene |
Show More |
Safety and Handling
Product Identifier
- Anthranilamide
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'