missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 39-122 Series Nitrile Heavy Duty Gloves
81.12 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
8
9
10
Description
- Liquid and chemical protection gloves
- Industrial gloves
- Tough glove offers optimum comfort of use thanks to its natural shape that makes it well suited for use with long-lasting tasks
- Superior dexterity and the high levels of wet and dry grip contribute to wearer safety and ultimately to the overall productivity
- Rugged glove with chemical protection
Applications:
- Adjusting panels parts components
- Applying finish to materials, products
- Assembly and disassembly of equipment
- Equipment repair, maintenance
- Handling materials with sharp or rough edges
- Oil, fluids and filter change
- Production line support and maintenance
Specifications
Certifications/Compliance | FDA 21 CFR 177.2600 |
Chlorinated | No |
Color | Green |
Material | Nitrile |
Size | 9 |
For Use With (Application) | Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work |
Glove Cuff Style | Gauntlet |
Lining | Lined |
Model | 39-122 9 217801 |
Series | AlphaTec™ 39-122 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'