Learn More
Ansell™ KSR™ 22-515 Series Vinyl-Coated Light Weight Gloves
KSR™ 22-515 Series Gloves - Light, flexible and comfortable. Ideal for long-wearing light-duty work. Excellent grip for handling small and difficult objects.
Brand: Ansell™ 22515080
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Ideal for general-purpose applications, as well as shipping, receiving, assembly, inspection, packaging, and maintenance.
- Last at least six times longer than 8 oz. (227g) cotton gloves
- Interlock knit lining stretches to provide snug, sensitive fit and ensures easy donning and doffing
- Won't shrink, stiffen, or lose grip; lining won't ball up
- Liquid repellent and machine washable
Specifications
Vinyl | |
Medium |