missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ KSR™ 22-515 Series Vinyl-Coated Light Weight Gloves

KSR™ 22-515 Series Gloves - Light, flexible and comfortable. Ideal for long-wearing light-duty work. Excellent grip for handling small and difficult objects.

Brand:  Ansell™ 22515090

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12630782

  • £53.12 / 12 pairs


Estimated Shipment
04-05-2021
Add to basket

Description

Description

Ideal for general-purpose applications, as well as shipping, receiving, assembly, inspection, packaging, and maintenance.

  • Last at least six times longer than 8 oz. (227g) cotton gloves
  • Interlock knit lining stretches to provide snug, sensitive fit and ensures easy donning and doffing
  • Won't shrink, stiffen, or lose grip; lining won't ball up
  • Liquid repellent and machine washable

Specifications

Specifications

Textured
Launderable
Slip-on
9
Tan
assembling, inspecting and packing small or delicate parts
Vinyl
24.5 cm
Videos
SDS
Documents