Learn More
Ansell ActivArmr™ 78-101 Seamless Knit Thermal Gloves
ActivArmr™ gloves dry faster than cotton to efficiently eliminate moisture from the skin. Special hollow-core fibers provide an effective thermal insulation barrier between the skin and surrounding environment.
Brand: Ansell 78101070
Code : S4
Additional Details : Weight : 0.36000kg
Description
- Gloves dry faster than cotton to efficiently eliminate moisture from the skin
- Special hollow-core fibres provide a thermal insulation barrier between the skin and surrounding environment
- Automatic knit technology construction for longer, more comfortable wear
- Seamless design eliminates any potential discomfort or skin irritation
- Thermal insulation for cold weather protection
- Ambidextrous to fit either hand
Specifications
ANSI 3 | |
Blue | |
Thermolite | |
214XA | |
Knit | |
13 |
EN 420:2003 + A1:2009; Category III | |
Ambidextrous | |
7 | |
Automotive Assembly, Food Processing | |
78-101 |