missing translation for 'onlineSavingsMsg'
Learn More

Ansell Hynit™ 32-125 Nitrile Coated Liquid Repellent Gloves

Catalog Number 12630832
Click to view available options
Size:
7
7.5
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Special nitrile-coating formulation is flexible while also repelling liquid, oil, and dirt. Gloves are compliant with FDA Food Handling Requirements.

  • Liquid-repellent impregnation repels oil, grease, and dirt
  • Nitrile coating is flexible for optimum dexterity
  • Compliant with FDA food handling requirements
  • Resistant to abrasions
ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Product Type Dip-coated Gloves
Size 7
For Use With (Application) Automotive Assembly, Equipment Operation, Metal Fabrication, and Mining
Glove Cuff Style Open
Liner Material Cotton/Polyester Blend
Packaging Type Polybag
Series 32-125
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'