Learn More
Ansell™ HyFlex™ 11-801 Foam Nitrile-Coated Gloves
These nitrile foam gloves offer an improved design that delivers up to 30% more breathability. For all day, every day comfort.
Brand: Ansell™ 11801110
Code : NEW
Additional Details : Weight : 0.01000kg
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
- Knit nylon liner enhances comfort
- Ultralight HyFlex glove offers a high degree of dexterity
- Ideal for general assembly and working with small, oily parts
- Reduce hand fatigue while increasing productivity
- Available with knitted variable stitch design; provides exceptional comfort
Compliance
Meets ANSI A1 (cut level) and ANSI 3 (abrasion level); EN 388-1149 standards.
Specifications
Palm | |
assembling, inspecting and packing small or delicate parts | |
Nylon | |
11 | |
20 to 26.5 cm |
Knit | |
Nitrile | |
11-801 | |
Black, Gray |