Learn More
Ansell™ HyFlex™ 11-800 Foam Nitrile-Coated Gloves
Foam nitrile coating is highly durable, resisting snags and abrasion. By channeling oil away from the glove's surface, the foam coating provides an excellent grip.
Brand: Ansell™ 11800060
Code : S4
Additional Details : Weight : 0.31500kg
Alert:
Due to extraordinary demand patterns, this item may experience longer lead times and shipping/delivery dates may not be accurate.
Description
- Knit nylon liner enhances comfort
- Ultralight HyFlex glove offers a high degree of dexterity
- Ideal for general assembly and working with small, oily parts
- Reduce hand fatigue while increasing productivity
- Available with knitted variable stitch design; provides exceptional comfort
Compliance
Meets ANSI A1 (cut level) and ANSI 3 (abrasion level); EN 388-1149 standards.
Specifications
Palm | |
Palm coated | |
General assembly and working with small, oily parts | |
Nylon | |
6 | |
Gray, White |
ZONZ™ Comfort Fit technology | |
Elastic knit | |
Foam Nitrile | |
11-800 | |
Industrial gloves | |
20 to 26.5 cm |