Learn More
Ansell™ HyFlex™ CR Plus Foam-Nitrile-Dipped Work Gloves
Kevlar™ Stretch Armor construction combines superior cut resistance and outstanding dexterity
Brand:  Ansell™ 11501080
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12610702
  • £204.27 / 12 pairs
Estimated Shipment
27-02-2018
Add to basket
Specifications
Specifications
Palm
Metal fabrication, body assembly, press and stamping, welding Glass manufacturing, Plastics industry
Kevlar™
Gray
Elastic knit
Nitrile foam
8
20.3 to 25.5cm
Videos