missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 11-501 Aramid Fiber Stretch Armor Gloves, Nitrile Coated

Catalog Number p-3080778
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

Ergonomic design reduces hand fatigue and provides a fit that mimics bare hands. Nitrile foam coating enhances tactile sensitivity and grip capabilities in dry or light oil conditions.

  • Intercept Technology™ raises the protection & dexterity standards for cut-resistant gloves
  • Aramid fiber provides an additional level of protection
  • Ergonomic design provides bare-hand closeness and reduces hand fatigue
  • Nitrile foam coating enhances grip capabilities and tactile sensitivity
ANSI Abrasion Rating ANSI 3
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A5
Hand Specific
Material Aramid Fiber
Product Type Cut Resistant Gloves
Size 6
For Use With (Application) Automotive Repairs, Machinery/Equipment Operation, Metal Fabrication, and Chemical Handling
Glove Cuff Style Knit
Series 11-501
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'