missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Hycron™ Nitrile-Coated Gloves

Made extra tough for heavy-duty jobs involving rough, abrasive materials. Ansell Hycron™ Nitrile-Coated Gloves have nitrile dipped coating on jersey liner.

Brand:  Ansell™ 27600080

Code : S4

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12630792

  • £36.00 / 12 pairs


In Stock 10
Add to basket

Description

Description

Available in three styles: nitrile-coated palm with knitwrist, full nitrile coating with knitwrist, and nitrile-coated palm with safety cuff. Nitrile coating ensures excellent handling of rough, abrasive materials while providing superior resistance to cuts, snags, punctures, abrasion, grease, and oil.

  • Two-piece jersey lining eliminates seams from working areas to prevent irritation and premature wear-through
  • Curved, preflexed fingers and wing thumb offer maximum worker comfort
  • Ideal for handling heavy-duty castings, cores, fabricated metal, concrete block, brick, tile, lumber, drywall, and plywood

Specifications

Specifications

Construction: Cut and sewn
Knitwrist
Nitrile
Cotton Jersey
8
EN 420:2003 + A1:2009 and EN388:2003
Industrial gloves
246 to 272mm
3/4 Dipped
Yes
12 pairs in a bag; 12 bags in a carton
27-600
40°C
Nitrile gloves
Blue
Videos