Learn More
Ansell™ AlphaTec™ 58-530B Series Nitrile Immersion Gloves
Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.
Brand: Ansell 58530B090
Code : S4
Additional Details : Weight : 0.54000kg
Description
- Liquid and chemical protection gloves
- Incorporates revolutionary Ansell Grip Technology™ to enable users to handle wet or oily objects with less grip force and more control
- Unique combination of liquid-proof chemical resistance and grip, together with flexibility and dexterity, makes the logical choice when working with chemicals
- Reliable chemical protection for added user safety
- Patented Ansell Grip Technology™ ensures that the coating polymer does not penetrate the liner during manufacture
- Coating is therefore consistent and sound, significantly reducing the likelihood of chemical leakage onto the skin
- AlphaTec™ models with white nylon liner are recommended for indoor applications, whereas with black acrylic liner are recommended for outdoor applications
Applications:
- Applying sealers, touching up
- Body painting including surface inspection
- Changing tools and dies
- Handling application and cleaning tools
- Loading and unloading process equipment
- Metalworking and assembly operations
- Opening furnaces draining pumps valves or lines and crackers BTX process
- Painting
- Painting or cleaning tools or robots
- Transferring liquids and solids between vessels and tanks and process equipment
- Warehousing of raw materials
Specifications
FDA 21 CFR 177-2600 | |
Nitrile | |
13 mil | |
Gauntlet | |
30.5 cm |
Black, Red | |
9 | |
Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work | |
12 in. |