Learn More
Ansell AlphaTec™ 58-535B Series Nitrile Immersion Gloves
Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.
Brand: Ansell 58535B100
43.84 GBP valid until 2024-03-30
Use promo code "21677" to get your promotional price.
Code : S4
Additional Details : Weight : 0.66240kg
Description
Provides dexterity with resistance to cuts, punctures, and abrasions as well as petrochemicals, oils, greases, pesticides, and other agricultural chemicals- Chemical resistant
- Material fully coated nitrile gloves
- Thickness: 11 mil
- Embossed grip surface for superior wet and oil grip
- Red with black coating
- Comfortable design reduces fatigue
Specifications
FDA 21 CFR 177-2600 | |
Yes | |
10 | |
0.33 mm | |
Gauntlet | |
58-535 | |
35.5 cm |
Green, Black | |
Nitrile | |
13 mil | |
Chemical plants and transportation, chemical processing, oil refining and petrochemicals, cleaning and de-greasing, exposure to chemicals, solvents, hazardous liquids, PVC cements or standing water, janitorial and maintenance work | |
Knit Lined Acrylic | |
14 in. |