Ansell™ 73-500 Touch N Tuff™ Sensitive Neoprene Gloves
Provides chemical splash resistance against wide variety of chemicals
Overview
Brand: Ansell™
Manufacturer Part Number: 73-500_8
Additional Details:
Additional Details: Weight: 0.28000kg
Product Code. 15324755
 N/A
N/A
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
  • Description and Specification
  • Alternative Products
Description and Specification
Specification
Glove Cuff Style Long, beaded
Hand Anatomic
For Use With (Application) Ideal for double donning and dry applications
Latex Free Yes
Material Neoprene
Size 8
Width (Metric) 101mm
Certifications CE, ISO 9001, ISO 13485 and ISO 14001
Alternative Products