Learn More
ANR49 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565499
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: KGNDDGIPDQE NSLDFSEHFN QLELLETHGH LIPTGTQSLW VGNSDEDEEQ DDKNEEWYRL QEKKMEKDPS RLLLWAAE Highest antigen sequence identity to the following orthologs - mouse 84%, rat 78%.
ANR49 Polyclonal antibody specifically detects ANR49 in Human samples. It is validated for Immunocytochemistry.Specifications
ANR49 | |
Polyclonal | |
Unconjugated | |
ANKRD49 | |
ANKRD49; ankyrin repeat domain 49; ankyrin repeat domain-containing protein 49; AW060934; C86108; fetal globin inducing factor; fetal globin-increasing factor; Fetal globin-inducing factor; FGIF; Gbif; globin inducing factor, fetal | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVL7 | |
ANKRD49 | |
Recombinant Human ANR49 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only