Learn More
Anpirtoline hydrochloride, Tocris Bioscience™
Highly potent 5-HT1B agonist; also 5-HT3 antagonist
Brand:  Tocris (Bio-Techne) 070310
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13437883
  • £151.80 / 10mg
Estimated Shipment
09-07-2019
Description
Description
Highly potent 5-HT1B receptor agonist (Ki values are 28, 150 and 1490 nM at 5-HT1B, 5-HT1A and 5-HT2 receptors respectively). Decreases central serotonin synthesis and attenuates aggressive behavior in vivo. Also acts as an antagonist at 5-HT3 receptors (Ki = 29.5 nM) and is brain penetrant.
Specifications
Chemical Identifiers
99201-87-3
265.196
anpirtoline hydrochloride, 2-chloro-6-piperidin-4-ylthio pyridine hydrochloride, 2-chloro-6-piperidin-4-ylsulfanylpyridine hydrochloride, pyridine, 2-chloro-6-4-piperidinylthio-, monohydrochloride, 2-chloro-6-piperidin-4-ylsulfanyl 2,6-14c2 pyridine hydrochloride 1:1, 2-chloro-6-piperidin-4-ylsulfanyl pyridine, hydrochloride, 2-chloro-6-4-piperidinylthio pyridine hydrochloride, 2-chloro-6-piperidin-4-ylsulfanyl pyridine hydrochloride
2-chloro-6-piperidin-4-ylsulfanylpyridine;hydrochloride
C10H14Cl2N2S
GRXDJABVNGUGCW-UHFFFAOYSA-N
6917943
C1CNCCC1SC2=NC(=CC=C2)Cl.Cl
Specifications
Anpirtoline hydrochloride
99201-87-3
anpirtoline hydrochloride, 2-chloro-6-piperidin-4-ylthio pyridine hydrochloride, 2-chloro-6-piperidin-4-ylsulfanylpyridine hydrochloride, pyridine, 2-chloro-6-4-piperidinylthio-, monohydrochloride, 2-chloro-6-piperidin-4-ylsulfanyl 2,6-14c2 pyridine hydrochloride 1:1, 2-chloro-6-piperidin-4-ylsulfanyl pyridine, hydrochloride, 2-chloro-6-4-piperidinylthio pyridine hydrochloride, 2-chloro-6-piperidin-4-ylsulfanyl pyridine hydrochloride
C1CNCCC1SC2=NC(=CC=C2)Cl.Cl
265.196
265.2
10mg
C10H14Cl2N2S
GRXDJABVNGUGCW-UHFFFAOYSA-N
2-chloro-6-piperidin-4-ylsulfanylpyridine;hydrochloride
6917943
>99%
Videos