missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Annexin V-FITC Apoptosis Detection Kit, for in vitro diagnostic use

Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry.

Brand:  Invitrogen™ BMS500FI/100CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15591947

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Includes: Annexin V-FITC protein
Binding Buffer (4x)
Propidium Iodide (20 ug/mL)

Description

Description

  • Blue Laser
  • Excitation at 488nm
  • Emission at 520nm
Specifications

Specifications

100 tests
Flow Cytometric Analysis
35.8 kd
FITC
>98%
Lipocortin V, Endonexin II, Calphobindin I
Videos
SDS
Documents

For Research Use Only