missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Annexin V-FITC Apoptosis Detection Kit

Annexin V staining, paired with 7-AAD or PI is widely used to identify apoptotic stages by flow cytometry.

Brand:  Invitrogen™ BMS500FI-300

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15521957

  • £287.48 / 300 tests


EU Stock 30
Add to basket

Disclaimers:

For Research Use Only


Includes: Annexin V-FITC protein
Binding Buffer (4x)
Propidium Iodide (20 ug/mL)

Description

Description

  • Blue Laser
  • Excitation at 488nm
  • Emission at 520nm
Specifications

Specifications

300 tests
Flow Cytometric Analysis
35.8 kd
FITC
> 98%
Lipocortin V, Endonexin II, Calphobindin I
Videos