missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kit PerCP-eFluor™ 710

Pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10 μM Camptothecin for 4 hours. This can be used at 5μL per test.

Brand:  Invitrogen™ 88-8008-72

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15591187

  • £138.00 / 50 tests

EU Stock 2

Please to purchase this item. Need a web account? Register with us today!

Alert:

Contains Sodium azide


Includes: Cat. No. 88-8008-72:
10X Binding Buffer
Annexin V PerCP-eFluor™ 710 : 5μL/test, 50 tests Protect from light.
Cat. No. 88-8008-74:
10X Binding Buffer
Annexin V PerCP-eFluor™ 710 : 5μL/test, 200 tests Protect from light.

Description

Description

Fixable Viability Dye eFluor™ 450 is not recommended for use with Annexin V Apoptosis Detection Kits.
Specifications

Specifications

50 tests
Flow Cytometric Analysis
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2° to 8°C. Do not freeze. Light sensitive material.
Videos
Documents

For Research Use Only