missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Annexin V Apoptosis Detection Kit eFluor™ 450

Compatible with intracellular staining. Pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10 μM Camptothecin for 4 hours. Both the Annexin V eFluor 450 and the 7-AAD components can be used at 5μL per test.

Brand:  Invitrogen™ 88-8006-72

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15561187

  • £123.00 / 50 tests

Estimated Shipment date
from Supplier 26-02-2020

Please to purchase this item. Need a web account? Register with us today!

Alert:

Contains Sodium azide


Includes: Cat. No. 88-8006-72:
10X Binding Buffer
Annexin V eFluor™ 450 : 5μL/test, 50 tests Protect from light.
7-AAD Viability Staining Solution : 5μL/test, 100 tests Protect from light.
Cat. No. 88-8006-74:
10X Binding Buffer
Annexin V eFluor™ 450 : 5μL/test, 200 tests Protect from light.
7-AAD Viability Staining Solution : 5μL/test, 2x100 tests Protect from light.

Description

Description

The components in this kit have been pre-titrated and tested on mouse thymocytes cultured overnight in medium (to induce apoptosis) or on Jurkat cells treated with 10μM Camptothecin for 4 hours. Both the Annexin V eFluor 450 and the 7-AAD components can be used at 5μL per test and should be protected from light.

Annexins are a family of calcium-dependent phospholipid-binding proteins that preferentially bind phosphatidylserine (PS). Under normal physiologic conditions, PS is predominantly located in the inner leaflet of the plasma membrane. Upon initiation of apoptosis, PS loses its asymmetric distribution across the phospholipid bilayer and is translocated to the extracellular membrane leaflet marking cells as targets of phagocytosis. Once on the outer surface of the membrane, PS can be detected by fluorescently labeled Annexin V in a calcium-dependent manner.

In early-stage apoptosis, the plasma membrane excludes viability dyes such as propidium iodide (PI), 7-AAD, or Fixable Viability Dyes such as eFluor™ 660 or eFluor™ 780. These cells will stain with Annexin V but not a viability dye, thus distinguishing cells in early apoptosis. However, in late stage apoptosis, the cell membrane loses integrity thereby allowing Annexin V to also access PS in the interior of the cell. A viability dye can be used to resolve these late-stage apoptotic and necrotic cells (Annexin V, viability dye-positive) from the early-stage apoptotic cells (Annexin V positive, viability dye-negative).

Specifications

Specifications

50 tests
Flow Cytometric Analysis
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
2° to 8°C. Do not freeze. Light sensitive material.
Videos
Documents

For Research Use Only