missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human M-CSF, Recombinant Protein, Carrier-Free

Human Recombinant Protein

Brand:  Invitrogen™ 34-8789-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15528836

  • £390.89 / 100µg


EU Stock 1
Add to basket

Description

Description

Human Macrophage Colony Stimulating Factor (M-CSF, also called CSF-1) is a ∽36 kDa disulfide-linked homodimer produced by a variety of cells including fibroblasts, macrophages, bone marrow stromal cells and endothelial cells. M-CSF regulates macrophage survival, proliferation and differentiation. M-CSF also enhances macrophage killing of tumor cells and microorganisms and regulates cytokine production from macrophages.
Specifications

Specifications

NM-172211
Sterile liquid: phosphate-buffered saline, 0.22μm filtered
M-CSF
>95%
Insect Cells
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
The bioactivity of this protein, as determined in a cell proliferation assay on M-NFS-60 cells, is 3ng/mL. This corresponds to a specific activity of 3.3 X 105 Units/mg.
Unlabeled
Yes
Cytokine Bioassays
36kDa
SDS-PAGE
100μg
Store at less than or equal to -70°C.
Human M-CSF Glu34-Ser190 accession number NM-172211 expressed in insect cells with a C-terminal 6X His tag.
CSF1, Macrophage colony-stimulating factor 1
Macrophage colony stimulating factor, monocyte colony stimulating factor
Recombinant Protein, Carrier-Free
Human
Videos