missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A8/ANXA8 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97052CL1
This item is not returnable.
View return policy
Description
Annexin A8/ANXA8 Polyclonal antibody specifically detects Annexin A8/ANXA8 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Annexin A8/ANXA8 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| annexin A8, Annexin VIII, Annexin-8, ANX8, FLJ53095, VAC-beta, Vascular anticoagulant-beta | |
| Produced in rabbits immunized with purified, recombinant Human Annexin A8/ANXA8 (Accession#: AAH73755.1; Met1-Pro327). | |
| 0.1 mL | |
| Signal Transduction | |
| 653145 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction