missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD63 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549152
Additional Details : Weight : 0.01000kg
Description
ANKRD63 Polyclonal antibody specifically detects ANKRD63 in Human samples. It is validated for Western Blot.Specifications
ANKRD63 | |
Polyclonal | |
Unconjugated | |
ANKRD63 | |
ANKRD63; ankyrin repeat domain 63; ankyrin repeat domain-containing protein 63 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100131244 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
C9JTQ0 | |
ANKRD63 | |
KLH conjugated synthetic peptide between 229-255 amino acids from the Central region of human ANKRD63. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |