missing translation for 'onlineSavingsMsg'
Learn More

ANKRD10 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA558577

Additional Details : Weight : 0.02000kg

Product Code. 15776451

  • £427.00 / 100µL
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: DCEGETPIHK AARSGSLECI SALVANGAHV DLRNASGLTA ADIAQTQGFQ ECAQFLLNLQ NCHLNHFYNN GILNGGHQNV FPNHISVGTN RKRCLEDSE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 81%.

ANKRD10 Polyclonal antibody specifically detects ANKRD10 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Specifications

Specifications

ANKRD10
Polyclonal
Unconjugated
ANKRD10
4833425P12Rik; ANKRD10; ankyrin repeat domain 10; ankyrin repeat domain-containing protein 10; AW549277
Rabbit
Antigen affinity chromatography
RUO
55608
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NXR5
ANKRD10
Recombinant protein corresponding to Human ANKRD10
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only