Learn More
ANKFY1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566801
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: ICTRGADMSVP DEKGNPPLWL ALANNLEDIA STLVRHGCDA TCWGPGPGGC LQTLLHRAID ENNEPTACFL IRSGCDVNSP R Highest antigen sequence identity to the following orthologs - mouse 95%, rat 96%.
ANKFY1 encodes a cytoplasmic protein that contains a coiled-coil structure and a BTB/POZ domain at its N-terminus, ankyrin repeats in the middle portion, and a FYVE-finger motif at its C-terminus. This protein belongs to a subgroup of double zinc finger proteins which may be involved in vesicle or protein transport.Specifications
ANKFY1 | |
Polyclonal | |
Unconjugated | |
ANKFY1 | |
Ankfy1; ANKHZN; ankyrin repeat and FYVE domain containing 1; ankyrin repeat and FYVE domain-containing protein 1; ankyrin repeat hooked to a zinc finger motif; ankyrin repeat hooked to zinc finger motif; ankyrin repeats hooked to a zinc finger motif; BTBD23; KIAA1255; mKIAA1255; rabankyrin-5; Rank-5; ZFYVE14 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51479 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P2R3 | |
ANKFY1 | |
Recombinant Human ANKFY1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only