Learn More
ANKAR Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557553
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QLPPAYYDTR IGQILINIDY MLKALWHGIY MPKEKRARFS ELWRAIMDID PDGKPQTNKD IFSEFSSAGL TDITKDPDFN EIYDEDVNED P Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.
ANKAR Polyclonal antibody specifically detects ANKAR in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
ANKAR | |
Polyclonal | |
Unconjugated | |
ANKAR | |
ANKAR; ankyrin and armadillo repeat containing; ankyrin and armadillo repeat-containing protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150709 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z5J8 | |
ANKAR | |
Recombinant protein corresponding to Human ANKAR | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only