missing translation for 'onlineSavingsMsg'
Learn More

Anisomycin, Tocris Bioscience™

JNK, SAPK and p38 activator

Brand:  Tocris (Bio-Techne) 129050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13469063

  • £358.68 / 50mg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Protein synthesis inhibitor (blocks translation). Potent activator of stress-activated protein kinases (JNK/SAPK) and p38 MAP kinase. Acts as a potent signaling agonist to selectively elicit homologous desensitization of immediate early gene induction (c-fos, fosB, c-jun, junB and junD).
Specifications

Chemical Identifiers

22862-76-6
265.309
(2R,3S,4S)-2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate
CHEBI:338412
CC(=O)OC1C(CNC1CC2=CC=C(C=C2)OC)O
C14H19NO4
YKJYKKNCCRKFSL-RDBSUJKOSA-N
253602
[(2R,3S,4S)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl] acetate

Specifications

Anisomycin
C14H19NO4
YKJYKKNCCRKFSL-RDBSUJKOSA-N
[(2R,3S,4S)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl] acetate
253602
265.31
50mg
22862-76-6
(2R,3S,4S)-2-[(4-Methoxyphenyl)methyl]-3,4-pyrrolidinediol 3-acetate
CC(=O)OC1C(CNC1CC2=CC=C(C=C2)OC)O
265.309
CHEBI:338412
>98%
Videos
Documents