Learn More
Aniracetam, Tocris Bioscience™
Inhibits desensitization; AMPA > kainate
Brand:  Tocris (Bio-Techne) 0867/50
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13428083
  • £111.72 / 50mg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Nootropic, with modulatory actions through allosteric potentiation of AMPA specific receptors, reduction of glutamate receptor desensitization and potentiation of metabotropic glutamate receptor activity. Anxiolytic following systemic administration.
Specifications
Chemical Identifiers
72432-10-1
219.24
aniracetam, draganon, sarpul, 1-4-methoxybenzoyl-2-pyrrolidinone, ampamet, 1-4-methoxybenzoyl pyrrolidin-2-one, 1-p-anisoyl-2-pyrrolidinone, memodrin, aniracetamun inn-latin, aniracetam usan:inn:jan
CHEBI:47943
COC1=CC=C(C=C1)C(=O)N2CCCC2=O
C12H13NO3
ZXNRTKGTQJPIJK-UHFFFAOYSA-N
2196
1-(4-methoxybenzoyl)pyrrolidin-2-one
Specifications
50mg
72432-10-1
aniracetam, draganon, sarpul, 1-4-methoxybenzoyl-2-pyrrolidinone, ampamet, 1-4-methoxybenzoyl pyrrolidin-2-one, 1-p-anisoyl-2-pyrrolidinone, memodrin, aniracetamun inn-latin, aniracetam usan:inn:jan
COC1=CC=C(C=C1)C(=O)N2CCCC2=O
219.24
CHEBI:47943
>99%
Aniracetam
C12H13NO3
ZXNRTKGTQJPIJK-UHFFFAOYSA-N
1-(4-methoxybenzoyl)pyrrolidin-2-one
2196
219.24