missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Anilinoacetaldehyde diethyl acetal, 98%

Brand:  Alfa Aesar™ A11587.06

Code : NEW

Additional Details : CAS Number : 22758-34-5 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 11424663

  • £60.95 / 5g


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

Anilinoacetaldehyde diethyl acetal
C6H5NHCH2CH(OCH2CH3)2
MFCD00051601
N-Phenylaminoacetaldehyde diethyl acetal
22758-34-5
209.29
2806325

Specifications

Anilinoacetaldehyde diethyl acetal
98%
209.29
2806325
153° to 154°C (12mmHg)
22758-34-5
C6H5NHCH2CH(OCH2CH3)2
MFCD00051601
N-Phenylaminoacetaldehyde diethyl acetal
5g
Videos
SDS