missing translation for 'onlineSavingsMsg'
Learn More

Aniline, 99.8%, pure, Thermo Scientific Chemicals

Catalog Number p-3735537
17.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


14.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 mL
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62-53-3
C6H7N
93.13
MFCD00007629
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
6115
CHEBI:17296
aniline
NC1=CC=CC=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
62-53-3
C6H7N
93.13
MFCD00007629
PAYRUJLWNCNPSJ-UHFFFAOYSA-N
benzenamine, phenylamine, aminobenzene, aminophen, anilin, kyanol, arylamine, cyanol, benzeneamine, benzidam
6115
CHEBI:17296
aniline
NC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -6.2°C
Color Pink to White
Density 1.0213g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Linear Formula C6H5NH2
Refractive Index 1.5845 to 1.5865
Quantity 5 mL
Beilstein 12, 59
Fieser 06,21; 17,16
Merck Index 15, 652
Specific Gravity 1.0213
Solubility Information Solubility in water: 36g/L (20°C).
Viscosity 4.4 mPa.s (20°C)
Formula Weight 93.13
Vapor Pressure 0.5mmHg at 20°C
Percent Purity 99.8%
Grade Pure
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material Aniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'