missing translation for 'onlineSavingsMsg'
Learn More

Sigma Laborzentrifugen™ Angle Rotors for Sigma™ Series 3 Centrifuges

Use these rotors with maximum load and at maximum speed for up to 50,000 cycles with low maintenance. Sigma™ Angle Rotors for Sigma™ Series 3 Centrifuges are ideal for use in research laboratories.

Brand:  Sigma Laborzentrifugen™ 12150

Code : NEW

Additional Details : Weight : 2.50000kg

 View more versions of this product

Product Code. 10056141

  • £1155.00 / Each

Estimated Shipment
12-05-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

Angle Rotor
21,000rpm
25°
6 x 50mL
Sigma 3-30KS, 3-16, 3-16K, 3-18K/KH
Videos
Documents