missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiostatin Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
POA
Specifications
Antigen | Angiostatin |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Angiostatin Polyclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.Specifications
Angiostatin | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
5340 | |
E. coli-derived recombinant human Angiostatin Kringle 1-3 Val98-Ser358 with Cys188Ser & Cys316Ser substitutions Accession # P00747 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Angiostatin in direct ELISAs and Western blots. This antibody will also recognize human plasminogen. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P00747 | |
PLG | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only