Learn More
Angiostatin Kringle 1-3 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF226
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447444
  • £362.60 / 100µg
Estimated Shipment
13-08-2020
Description
Description
Angiostatin Kringle 1-3 Polyclonal antibody AF226 Specifically detects Angiostatin Kringle 1-3 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Angiostatin
LYOPH
Western Blot 0.1 ug/mL
P00747
PLG
E. coli-derived recombinant human Angiostatin Kringle 1-3 Val98-Ser358 with Cys188Ser & Cys316Ser substitutions Accession # P00747
Immunogen affinity purified
RUO
Primary
5340
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Angiostatin in direct ELISAs and Western blots. This antibody will also recognize human plasminogen.