missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 3/ANGPTL3 Antibody, R&D Systems™
Sheep Polyclonal Antibody
POA
Specifications
Antigen | Angiopoietin-like Protein 3/ANGPTL3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Angiopoietin-like Protein 3/ANGPTL3 Polyclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot.Specifications
Angiopoietin-like Protein 3/ANGPTL3 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Sheep | |
Human | |
AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
ANGPTL3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9Y5C1 | |
27329 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Angiopoietin-like 3 (R&D Systems, Catalog # 3829-AN) Ser17-Glu460 Accession # Q9Y5C1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Angiopoietin-like 3 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Angiopoietin-like 3 is observed and less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-like 1 and rhAngiopoietin-like 4 is observed. |
For Research Use Only