missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Angiopoietin-like Protein 1/ ANGPTL1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | Angiopoietin-like Protein 1/ANGPTL1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Description
Angiopoietin-like Protein 1/ANGPTL1 Polyclonal specifically detects Angiopoietin-like Protein 1/ANGPTL1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Angiopoietin-like Protein 1/ANGPTL1 | |
Western Blot 0.1 ug/mL, Immunohistochemistry 3-15 ug/mL | |
Polyclonal | |
Goat | |
Human | |
ANG-3, ANG3angioarrestin, Angiopoietin like Protein 1, angiopoietin Y1, angiopoietin-3, angiopoietin-like 1, Angiopoietin-like protein 1, angiopoietin-related protein 1, ANGPT3, ANGPTL1, AngY, AngY1, ARP1, ARP1angiopoietin 3, dJ595C2.2, dJ595C2.2 (angiopoietin Y1), KIAA0351, UNQ162 | |
ANGPTL1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
RUO | |
O95841 | |
9068 | |
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-like 1/ANGPTL1 Ala39-Asp491 Accession # O95841 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Angiopoietin-like 1/ANGPTL1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) Angiopoietin-4 is observed and less than 5% cross-reactivity with rhAngiopietin-1 and rhAngiopoietin-2 is observed. |
For Research Use Only