missing translation for 'onlineSavingsMsg'
Learn More

Angiogenin Mouse anti-Human, Biotin, Clone: 26-2F, eBioscience™

Mouse Monoclonal Antibody

£55.60 - £141.00

Specifications

Antigen Angiogenin
Clone 26-2F
Host Species Murine
Gene Alias Ribonuclease 5
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Microscopy
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15567376 View Documents Affymetrix eBioscience
13-9762-80
25μg £55.60
25µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
15577376 View Documents Affymetrix eBioscience
13-9762-82
100μg £141.00
100µg View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 14-05-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The monoclonal antibody 26-2F recognizes human angiogenin. Angiogenin is a secreted ribonuclease (RNASE)-family member and is important for the formation of new blood vessels as well as the stimulation of cell proliferation and survival. Cellular stress signals direct angiogenin to stress granules where angiogenin functions by hydrolyzing cellular tRNA resulting in modulation of protein translation and promotion of cell survival. Cytoplasmic angiogenin can bind to actin, forming complexes that activate protease cascades which can degrade laminin and fibronectin allowing endothelial cells to penetrate the basement membrane and migrate. In cancer cells, growth signals direct angiogenin translocation to the nucleus where binding of DNA promotor regions stimulates rRNA transcription resulting in a downstream increase in protein synthesis driving cell growth and survival. Angiogenin is upregulated in multiple cancers including breast, prostate, cervical, colorectal, lymphoma, and melanoma. During development and in adulthood, angiogenin is highly expressed in the spinal cord and contributes to motor neuron survival. Angiogenin is important for neuronal survival and is depleted in neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS) and Parkinson’s disease. This 26-2F antibody is reported to have both in vivo and in vitro neutralizing activity to angiogenin.
Specifications

Specifications

Angiogenin
Murine
ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Microscopy
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
26-2F
Ribonuclease 5
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer

For Research Use Only.