missing translation for 'onlineSavingsMsg'
Learn More

Angiogenin Mouse anti-Human, Biotin, Clone: 26-2F, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-9762-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15577376

  • £141.00 / 100µg

Estimated Shipment
10-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The monoclonal antibody 26-2F recognizes human angiogenin. Angiogenin is a secreted ribonuclease (RNASE)-family member and is important for the formation of new blood vessels as well as the stimulation of cell proliferation and survival. Cellular stress signals direct angiogenin to stress granules where angiogenin functions by hydrolyzing cellular tRNA resulting in modulation of protein translation and promotion of cell survival. Cytoplasmic angiogenin can bind to actin, forming complexes that activate protease cascades which can degrade laminin and fibronectin allowing endothelial cells to penetrate the basement membrane and migrate. In cancer cells, growth signals direct angiogenin translocation to the nucleus where binding of DNA promotor regions stimulates rRNA transcription resulting in a downstream increase in protein synthesis driving cell growth and survival. Angiogenin is upregulated in multiple cancers including breast, prostate, cervical, colorectal, lymphoma, and melanoma. During development and in adulthood, angiogenin is highly expressed in the spinal cord and contributes to motor neuron survival. Angiogenin is important for neuronal survival and is depleted in neurodegenerative diseases such as amyotrophic lateral sclerosis (ALS) and Parkinson’s disease. This 26-2F antibody is reported to have both in vivo and in vitro neutralizing activity to angiogenin.
Specifications

Specifications

Angiogenin
26-2F
Biotin
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
ELISA, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Microscopy
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Ribonuclease 5
IgG1, kappa
RUO
Primary
Human
Videos
Documents

For Research Use Only.